Skip to Content
Merck
All Photos(2)

Key Documents

SAB1407805

Sigma-Aldrich

Anti-FANCM antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FAAP250, KIAA1596, MGC176453

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75.6 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FANCM(57697)

General description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group M. (provided by RefSeq)

Immunogen

FANCM (AAH36056.1, 1 a.a. ~ 669 a.a) full-length human protein.

Sequence
MSGRQRTLFQTWGSSISRSSGTPGCSSGTERPQSPGSSKAPLPAAAEAQLESDDDVLLVAAYEAERQLCLENGGFCTSAGALWIYPTNCPVRDYQLHISRAALFCNTLVCLPTGLGKTFIAAVVMYNFYRWFPSGKVVFMAPTKPLVTQQIEACYQVMGIPQSHMAEMTGSTQASTRKEIWCSKRVLFLTPQVMVNDLSRGACPAAEIKCLVIDEAHKALGNYAYCQVVRELVKYTNHFRILALSATPGSDIKAVQQVITNLLIGQIELRSEDSPDILTYSHERKVEKLIVPLGEELAAIQKTYIQILESFARSLIQRNVLMRRDIPNLTKYQIILARDQFRKNPSPNIVGIQQGIIEGEFAICISLYHGYELLQQMGMRSLYFFLCGIMDGTKGMTRSKNELGRNEDFMKLYNHLECMFARTRSTSANGISAIQQGDKNKKFVYSHPKLKKLEEVVIEHFKSWNAENTTEKKRDETRVMIFSSFRDSVQEIAEMLSQHQPIIRVMTFVGHASGKSTKGFTQKEQLEVVKQFRDGGYNTLVSTCVGEEGLDIGEVDLIICFDSQKSPIRLVQRMGRTGRKRQGRIVIILSEGREERIYNQSQSNKRSIYKAISSNRQVLHFYQRSPRMVPDGINPKLHKMFITHGVYEPEKPSRNLQRKSSIFSYRDGK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veselin Andreev et al.
DNA repair, 74, 38-50 (2019-01-05)
Chromatin regulators play crucial roles in the DNA damage response. While the chromatin changes needed for double-strand break repair and nucleotide excision repair have been intensely studied, the chromatin requirements of interstrand crosslink (ICL) repair have remained largely unexplored. Here
Laura Kasak et al.
American journal of human genetics, 103(2), 200-212 (2018-08-04)
Infertility affects around 7% of men worldwide. Idiopathic non-obstructive azoospermia (NOA) is defined as the absence of spermatozoa in the ejaculate due to failed spermatogenesis. There is a high probability that NOA is caused by rare genetic defects. In this
Jing Zhang et al.
Nature communications, 11(1), 3951-3951 (2020-08-10)
Duplication of mammalian genomes requires replisomes to overcome numerous impediments during passage through open (eu) and condensed (hetero) chromatin. Typically, studies of replication stress characterize mixed populations of challenged and unchallenged replication forks, averaged across S phase, and model a
Jing Huang et al.
Cell reports, 27(6), 1794-1808 (2019-05-09)
Eukaryotic replisomes are driven by the mini chromosome maintenance (MCM [M]) helicase complex, an offset ring locked around the template for leading strand synthesis by CDC45 (C) and GINS (G) proteins. Although the CDC45 MCM GINS (CMG) structure implies that
Daniel González-Acosta et al.
The EMBO journal, 40(14), e106355-e106355 (2021-06-16)
DNA interstrand crosslinks (ICLs) induced by endogenous aldehydes or chemotherapeutic agents interfere with essential processes such as replication and transcription. ICL recognition and repair by the Fanconi Anemia pathway require the formation of an X-shaped DNA structure that may arise

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service