Skip to Content
Merck
All Photos(4)

Key Documents

SAB1407335

Sigma-Aldrich

Anti-CACYBP antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

GIG5, MGC87971, PNAS-107, RP1-102G20.6, S100A6BP, SIP

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~26.2 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACYBP(27101)

General description

The protein encoded by this gene is a calcyclin binding protein. It may be involved in calcium-dependent ubiquitination and subsequent proteosomal degradation of target proteins. It probably serves as a molecular bridge in ubiquitin E3 complexes and participates in the ubiquitin-mediated degradation of beta-catenin. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CACYBP (NP_055227.1, 1 a.a. ~ 228 a.a) full-length human protein.

Sequence
MASEELQKDLEEVKVLLEKATRKRVRDALTAEKSKIETEIKNKMQQKSQKKAELLDNEKPAAVVAPITTGYTVKISNYGWDQSDKFVKIYITLTGVHQVPTENVQVHFTERSFDLLVKNLNGKSYSMIVNNLLKPISVEGSSKKVKTDTVLILCRKKVENTRWDYLTQVEKECKEKEKPSYDTETDPSEGLMNVLKKIYEDGDDDMKRTINKAWVESREKQAKGDTEF

Biochem/physiol Actions

CACYBP (calcyclin binding protein) is a part of the ubiquitination machinery and is involved in the degradation of β-catenin. It can interact with S100, SIAH1 (seven in absentia homolog 1) and SKP1 (S-phase kinase-associated protein 1). CACYBP is involved in proliferation, differentiation, multidrug resistance and tumorigenicity. It can work as an oncogene and as a tumor suppressor. In glioma cells, CACYBP enhances the cellular proliferation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CacyBP/SIP inhibits Doxourbicin-induced apoptosis of glioma cells due to activation of ERK1/2.
Tang Y, et al.
IUBMB Life, 68, 211-219 (2016)
CacyBP/SIP protein is important for the proliferation of human glioma cells.
Shi H, et al.
IUBMB Life, 66, 286-291 (2014)
Expression and regulation of CacyBP/SIP in chronic lymphocytic leukemia cell balances of cell proliferation with apoptosis.
Fu C, et al.
Journal of Cancer Research and Clinical Oncology, 142, 741-748 (2016)
S100A6 protein negatively regulates CacyBP/SIP-mediated inhibition of gastric cancer cell proliferation and tumorigenesis.
Ning X, et al.
PLoS ONE, 7, e30185-e30185 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service