Skip to Content
Merck
All Photos(1)

Documents

SAB1406786

Sigma-Aldrich

Anti-PCSK7 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

LPC, PC7, PC8, SPC7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~64.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCSK7(9159)

General description

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease. It is structurally related to its family members, PACE and PACE4. This protein is concentrated in the trans-Golgi network, associated with the membranes, and is not secreted. It can process proalbumin and is thought to be responsible for the activation of HIV envelope glycoproteins gp160 and gp140. This gene has been implicated in the transcriptional regulation of housekeeping genes. Multiple alternatively spliced transcripts are described for this gene but their full length nature is not yet known. Downstream of this gene′s map location at 11q23-q24, nucleotides that match part of this gene′s 3′ end are duplicated and inverted. A translocation breakpoint associated with lymphoma occurs between this gene and its inverted counterpart. (provided by RefSeq)

Immunogen

PCSK7 (AAH06357.1, 1 a.a. ~ 591 a.a) full-length human protein.

Sequence
MPKGRQKVPHLDAPLGLPTCLWLELAGLFLLVPWVMGLAGTGGPDGQGTGGPSWAVHLESLEGDGEEETLEQQADALAQAAGLVNAGRIGELQGHYLFVQPAGHRPALEVEAIRQQVEAVLAGHEAVRWHSEQRLLRRAKRSVHFNDPKYPQQWHLNNRRSPGRDINVTGVWERNVTGRGVTVVVVDDGVEHTIQDIAPNYSPEGSYDLNSNDPDPMPHPDVENGNHHGTRCAGEIAAVPNNSFCAVGVAYGSRIAGIRVLDGPLTDSMEAVAFNKHYQINDIYSCSWGPDDDGKTVDGPHQLGKAALQHGVIAGRQGFGSIFVVASGNGGQHNDNCNYDGYANSIYTVTIGAVDEEGRMPFYAEECASMLAVTFSGGDKMLRSIVTTDWDLQKGTGCTEGHTGTSAAAPLAAGMIALMLQVRPCLTWRDVQHIIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLELKLFCPSGMMSLIGAPRSMDSWLCVECSRHQGQTKAVRECHEWKIPAR

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service