Skip to Content
Merck
All Photos(1)

Documents

SAB1405888

Sigma-Aldrich

Anti-GPLD1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

GPIPLD, GPIPLDM, MGC22590, PIGPLD, PIGPLD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~19.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPLD1(2822)

General description

Many proteins are tethered to the extracellular face of eukaryotic plasma membranes by a glycosylphosphatidylinositol (GPI) anchor. The GPI-anchor is a glycolipid found on many blood cells. The protein encoded by this gene is a GPI degrading enzyme. Glycosylphosphatidylinositol specific phospholipase D1 hydrolyzes the inositol phosphate linkage in proteins anchored by phosphatidylinositol glycans, thereby releasing the attached protein from the plasma membrane. (provided by RefSeq)

Immunogen

GPLD1 (AAH20748.1, 1 a.a. ~ 176 a.a) full-length human protein.

Sequence
MSAFRLWPGLLIMLGSLCHRGSPCGLSTHIEIGHRALEFLQLHNGRVNYRELLLEHQDAYQAGIVFPDCFYPSICKGGKFHDVSESTHWTPFLNASVHYIRENYPLPWEKDTEKLVAFLFGITSHMAADVSWHSLGLEQGFLRTMGAIDFHGSYSEAHSAGDFGTVYLHLLNFLVV

Application

Anti-GPLD1 antibody produced in mouse is suitable for western blot analysis.

Biochem/physiol Actions

GPLD1 (Glycosylphosphatidylinositol specific phospholipase D1) is only expressed in human lver. Inositol phosphate linkage in proteins anchored by phosphatidylinositol-glycans (PI-Gs) is specifically hydrolyzed by GPLD1, suggesting GPLD1 expression might influence the location and expression of PI-G-anchored proteins. Studies have reported that it might be a significant factor in process of leukemia pathogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang-Fen Xiao et al.
Zhongguo shi yan xue ye xue za zhi, 18(1), 15-18 (2010-02-09)
This study was purposed to investigate the expression and significance of glycosylphosphatidylinositol-specific phospholipase D (GPI-PLD) in bone marrow mononuclear cells (BMMNC) isolated from patients with acute myeloid leukemia (AML), GPI-PLD activity in BMMNC isolated from 78 patients with AML and
B J Scallon et al.
Science (New York, N.Y.), 252(5004), 446-448 (1991-04-19)
A phosphatidylinositol-glycan-specific phospholipase D (PI-G PLD) that specifically hydrolyzes the inositol phosphate linkage in proteins anchored by phosphatidylinositol-glycans (PI-Gs) has recently been purified from human and bovine sera. The primary structure of bovine PI-G PLD has now been determined and
J N Schofield et al.
Biochimica et biophysica acta, 1494(1-2), 189-194 (2000-11-10)
Here we report the structure of the human glycosylphosphatidylinositol phospholipase D1 gene, which covers at least 80 kb on chromosome 6p22. The gene comprises 25 exons and encodes a 5.8 kb mRNA, which was detected only in the liver. Southern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service