Skip to Content
Merck
All Photos(2)

Key Documents

SAB1405470

Sigma-Aldrich

Anti-BIRC5 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

API4, EPR-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1591


Select a Size

Change View
50 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1591

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~16.4 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BIRC5(332)

General description

This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene′s expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. (provided by RefSeq)

Immunogen

BIRC5 (AAH08718.1, 1 a.a. ~ 142 a.a) full-length human protein.

Sequence
MGAPTLPPAWQPFLKDHRISTFKNWPFLEGCACTPERMAEAGFIHCPTENEPDLAQCFFCFKELEGWEPDDDPIEEHKKHSSGCAFLSVKKQFEELTLGEFLKLDRERAKNKIAKETNNKKKEFEETAKKVRRAIEQLAAMD

Application

Anti-BIRC5 antibody produced in mouse is suitable for indirect immunofluorescence and western blot applications.

Biochem/physiol Actions

BIRC5 (Baculoviral inhibitor of apoptosis repeat-containing 5) functions as a cell surface receptor for factor Xa. It is predicted to be a potential factor in protease-dependent cellular effector functions. It also behaves as a receptor for factor Xa during the cell surface assembly of proteolytic activities and leukocyte mitogenesis. A study reports that the expression of BIRC5 can be controlled by mRNA splicing. It is an inhibitor of apoptosis and is expressed in various malignancies.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaya Nigam et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(9), 9241-9246 (2014-06-18)
Survivin, an inhibitor of apoptosis, has been shown to be expressed in various malignancies. However, its role in gallbladder cancer (GBC) has not been evaluated yet. We investigated its expression in peripheral blood of patients with gallbladder diseases (gallstone disease
D C Altieri
The Journal of biological chemistry, 269(5), 3139-3142 (1994-02-04)
Cellular receptors for blood proteases regulate chemotaxis, extracellular proteolysis, and growth behavior of normal and malignant cells. Binding of the coagulation protease factor Xa to leukocytes is contributed by a recently identified molecule, denominated Effector cell Protease Receptor-1 (EPR-1). Monoclonal
D C Altieri
Biochemistry, 33(46), 13848-13855 (1994-11-22)
Effector cell protease receptor-1 (EPR-1) is a transmembrane glycoprotein receptor for factor Xa that contributes to cell surface assembly of proteolytic activities and leukocyte mitogenesis. It is now shown that membrane expression of EPR-1 is dynamically modulated by mRNA splicing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service