Skip to Content
Merck
All Photos(2)

Key Documents

SAB1405316

Sigma-Aldrich

Monoclonal Anti-TNS4 antibody produced in mouse

clone 1C1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CTEN, FLJ14950, PP14434

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~78.91 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNS4(84951)

General description

Mouse monoclonal antibody raised against a full-length recombinant TNS4.
TNS4 (Tensin 4) is a signaling molecule localized in the prostate and placenta. It is a member of the TENSIN gene family. It has a Src homology 2 and phosphotyrosine-binding domains. Its expression has been observed in the early progression of melanoma cells.

Immunogen

TNS4 (AAH18706, 1 a.a. ~ 480 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGSKASSPHGLGSPLVASPRLEKRLGGLAPQRGSRISVLSASPVSDVSYMFGSSQSLLHSSNSSHQSSSRSLESPANSSSSLHSLGSVSLCTRPSDFQAPRNPTLTMGQPRTPHSPPLAKEHASSCPPSITNSMVDIPIVLINGCPEPGSSPPQRTPGHQNSVQPGAASPSNPCPATRSNSQTLSDAPFTTCPEGPARDMQPTMKFVMDTSKYWFKPNITREQAIELLRKEEPGAFVIRDSSSYRGSFGLALKVQEVPASAQSRPGEDSNDLIRHFLIESSAKGVHLKGADEEPYFGSLSAFVCQHSIMALALPCKLTIPQRELGGADGASDSTDSPASCQKKSAGCHTLYLSSVSVETLTGALAVQKAISTTFERDILPTPTVVHFKVTEQGITLTDVQRKVFFRRHYPLTTLRFCGMDPEQRKWQKYCKPSWIFGFVAKSQTEPQENVCHLFAEYDMVQPASQVIGLVTALLQDAERM

Application

Monoclonal Anti-TNS4 antibody produced in mouse is suitable for indirect ELISA and western blot.

Biochem/physiol Actions

TNS4 (Tensin 4) is mainly involved in cell migration activity. 1 It exhibits oncogenic property in colonic, breast, lung, and gastric cancers. In studies, its overexpression has been reported in the pancreatic cancer cells. In pancreatic cancer cells, TNS4 promotes colony formation and cell motility. It has been predicted that TNS4 could be a prognostic marker for primary melanoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su Hao Lo et al.
Cancer research, 62(15), 4217-4221 (2002-08-03)
Tensin is a signaling molecule that binds to actin filaments and localizes to focal adhesions. Recently, we have discovered that tensin represents a new gene family with related members that are expressed in a variety of tissues. In this study
Cecilia Sjoestroem et al.
PloS one, 8(11), e80492-e80492 (2013-11-19)
C-terminal tensin-like protein (Cten) is a focal adhesion protein originally identified as a tumor suppressor in prostate cancer. It has since been found to be overexpressed and function as an oncogene in numerous other cancers, but the expression status of
Abdulkader Albasri et al.
Breast cancer research and treatment, 126(1), 47-54 (2010-04-15)
C-terminal tensin-like (CTEN) gene is a member of the TENSIN gene family, involved in cell migration and localised at focal adhesion sites. This study was designed to explore the prevalence and clinical significance of CTEN expression in a large and
Saleh Al-Ghamdi et al.
Pancreas, 42(1), 135-140 (2012-07-04)
C-terminal tensin-like gene (CTEN, also known as TNS4) localizes to focal adhesions and is reported to function as an oncogene in colonic, breast, lung, and gastric cancers. Its role in pancreatic cancer is unknown and was thus investigated in this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service