Skip to Content
Merck
All Photos(5)

Key Documents

SAB1404696

Sigma-Aldrich

Monoclonal Anti-MAPKAPK2 antibody produced in mouse

clone 3B8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MK2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.57 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPKAPK2(9261)

Related Categories

General description

Mitogen-activated protein kinase-activated protein kinase 2 (MAPKAPK2) gene encodes a member of the Ser/Thr protein kinase family. This kinase is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, this kinase is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for this gene. (provided by RefSeq)The gene is located on human chromosome 1q32.1. It consists of an autoinhibitory domain, a helix-turn-helix structure, which occupies the substrate binding cleft of the kinase domain and inhibits kinase function.

Immunogen

MAPKAPK2 (NP_116584, 266 a.a. ~ 352 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NHGLAISPGMKTRIRMGQYEFPNPEWSEVSEEVKMLIRNLLKTEPTQRMTITEFMNHPWIMQSTKVPQTPLHTSRVLKEDKERWEDV

Biochem/physiol Actions

Mitogen-activated protein kinase-activated protein kinase 2 (MAPKAPK2) is involved in cytokine production and cell migration. Overexpression of MAPKAPK2 confers multiple myeloma (MM) resistance to chemotherapy. It phosphorylates the proteins found in the nucleus and cytoplasm. This protein confers gemcitabine sensitivity in pancreatic cancer cells. The protein is required for tumor necrosis factor (TNF) biosynthesis. It is linked to tumorigenesis and drug resistance. The protein functions as a prognostic marker for lung cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A functional copy-number variation in MAPKAPK2 predicts risk and prognosis of lung cancer
Liu B, et al.
American Journal of Human Genetics, 91(2), 384-390 (2012)
MK2-TNF-Signaling Comes Full Circle
Menon MB, et al.
Trends in Biochemical Sciences (2017)
The MAPK-activated protein kinase 2 mediates gemcitabine sensitivity in pancreatic cancer cells
Kopper F, et al.
Cell Cycle, 13(6), 884-889 (2014)
MAPKAPK2 (mitogen-activated protein kinase-activated protein kinase 2)
Felix R, et al.
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service