Skip to Content
Merck
All Photos(1)

Key Documents

SAB1404664

Sigma-Aldrich

Monoclonal Anti-ANGPTL1, (C-terminal) antibody produced in mouse

clone 1C2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ANG3, ANGPT3, ARP1, AngY, KIAA0351, UNQ162, dJ595C2.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (c)
ELISA (i)
clone:
1C2, monoclonal
species reactivity:
human
citations:
technique(s):
capture ELISA: suitable
indirect ELISA: suitable

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~77.59 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANGPTL1(9068)

General description

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro. (provided by RefSeq)

Immunogen

ANGPTL1 (AAH50640, 24 a.a. ~ 491 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GQFKIKKINQRRYPRATDGKEEAKKCAYTFLVPEQRITGPICVNTKGQDASTIKDMITRMDLENLKDVLSRQKREIDVLQLVVDVDGNIVNEVKLLRKESRNMNSRVTQLYMQLLHEIIRKRDNSLELSQLENKILNVTTEMLKMATRYRELEVKYASLTDLVNNQSVMITLLEEQCLRIFSRQDTHVSPPLVQVVPQHIPNSQQYTPGLLGGNEIQRDPGYPRDLMPPPDLATSPTKSPFKIPPVTFINEGPFKDCQQAKEAGHSVSGIYMIKPENSNGPMQLWCENSLDPGGWTVIQKRTDGSVNFFRNWENYKKGFGNIDGEYWLGLENIYMLSNQDNYKLLIELEDWSDKKVYAEYSSFRLEPESEFYRLRLGTYQGNAGDSMMWHNGKQFTTLDRDKDMYAGNCAHFHKGGWWYNACAHSNLNGVWYRGGHYRSKHQDGIFWAEYRGGSYSLRAVQMMIKPID

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service