Skip to Content
Merck
All Photos(2)

Key Documents

SAB1404003

Sigma-Aldrich

Monoclonal Anti-KCNJ15 antibody produced in mouse

clone 1F8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

IRKK, KIR1.3, KIR4.2, MGC13584

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~67.36 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ15(3772)

General description

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein has a greater tendency to allow potassium to flow into a cell rather than out of a cell. Three transcript variants encoding the same protein have been found for this gene. (provided by RefSeq)

Immunogen

KCNJ15 (AAH13327, 1 a.a. ~ 375 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDAIHIGMSSTPLVKHTAGAGLKANRPRVMSKSGHSNVRIDKVDGIYLLYLQDLWTTVIDMKWRYKLTLFAATFVMTWFLFGVIYYAIAFIHGDLEPGEPISNHTPCIMKVDSLTGAFLFSLESQTTIGYGVRSITEECPHAIFLLVAQLVITTLIEIFITGTFLAKIARPKKRAETIKFSHCAVITKQNGKLCLVIQVANMRKSLLIQCQLSGKLLQTHVTKEGERILLNQATVKFHVDSSSESPFLILPMTFYHVLDETSPLRDLTPQNLKEKEFELVVLLNATVESTSAVCQSRTSYIPEEVYWGFEFVPVVSLSKNGKYVADFSQFEQIRKSPDCTFYCADSEKQQLEEKYRQEDQRERELRTLLLQQSNV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service