SAB1403984
Monoclonal Anti-INSRR antibody produced in mouse
clone 5C6, purified immunoglobulin, buffered aqueous solution
Synonym(s):
IRR
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
5C6, monoclonal
form
buffered aqueous solution
mol wt
antigen ~38.1 kDa
species reactivity
human, rat
technique(s)
capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... INSRR(3645)
General description
The gene INSRR (insulin receptor related receptor) is mapped to human chromosome 1q21-q23. It belongs to the insulin receptor family of proteins. INSRR is mainly present in organs which are associated with acid/base generation. The protein has two N-terminal leucine-rich repeat domains, a cysteine-rich C domain and three C-terminal fibronectin type-III repeats.
Immunogen
Sequence
LYLNDYCHRGLRLPTSNNDPRFDGEDGDPEAEMESDCCPCQHPPPGQVLPPLEAQEASFQKKFENFLHNAITIPISPWKVTSINKSPQRDSGRHRRAAGPLRLGGNSSDF
Biochem/physiol Actions
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service