Skip to Content
Merck
All Photos(3)

Key Documents

SAB1403896

Sigma-Aldrich

Monoclonal Anti-GTF3A antibody produced in mouse

clone 3A1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

AP2, TFIIIA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.01 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTF3A(2971)

General description

The product of this gene is a zinc finger protein with nine Cis[2]-His[2] zinc finger domains. It functions as an RNA polymerase III transcription factor to induce transcription of the 5S rRNA genes. The protein binds to a 50 bp internal promoter in the 5S genes called the internal control region (ICR), and nucleates formation of a stable preinitiation complex. This complex recruits the TFIIIC and TFIIIB transcription factors and RNA polymerase III to form the complete transcription complex. The protein is thought to be translated using a non-AUG translation initiation site in mammals based on sequence analysis, protein homology, and the size of the purified protein. (provided by RefSeq)

Immunogen

GTF3A (NP_002088, 185 a.a. ~ 274 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NQQKQYICSFEDCKKTFKKHQQLKIHQCQNTNEPLFKCTQEGCGKHFASPSKLKRHAKAHEGYVCQKGCSFVAKTWTELLKHVRETHKEE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service