Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403813

Sigma-Aldrich

Monoclonal Anti-FGF8 antibody produced in mouse

clone 2A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

AIGF, HBGF-8, MGC149376

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~33.6 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
proximity ligation assay: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF8(2253)

General description

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is known to be a factor that supports androgen and anchorage independent growth of mammary tumor cells. Overexpression of this gene has been shown to increase tumor growth and angiogensis. The adult expression of this gene is restricted to testes and ovaries. Temporal and spatial pattern of this gene expression suggests its function as an embryonic epithelial factor. Studies of the mouse and chick homologs revealed roles in midbrain and limb development, organogenesis, embryo gastrulation and left-right axis determination. The alternative splicing of this gene results in four transcript variants. (provided by RefSeq)

Immunogen

FGF8 (NP_149354, 65 a.a. ~ 133 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SRRLIRTYQLYSRTSGKHVQVLANKRINAMAEDGDPFAKLIVETDTFGSR VRVRGAETGLYICMNKKGK

Biochem/physiol Actions

Fibroblast growth factor-8 (FGF-8) stimulates the proliferation and activation of cells that express the FGF receptors. It has a role in embryogenesis. FGF8b, an isoform of FGF-8, is expressed in prostate cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening a phage display library for a novel FGF8b-binding peptide with anti-tumor effect on prostate cancer.
Wang W
Experimental Cell Research, 319(8), 1156-1164 (2013)
A novel locus of ectodermal dysplasia maps to chromosome 10q24.32-q25.1.
Rafiq MA
The Journal of Investigative Dermatology, 124(2), 338-342 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service