Skip to Content
Merck
All Photos(1)

Key Documents

SAB1403361

Sigma-Aldrich

Monoclonal Anti-LENG4 antibody produced in mouse

clone 1D4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

BB1, LENG4, LPIAT, hMBOA-7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.78 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBOAT7(79143)

General description

Membrane bound O-acyltransferase domain containing 7 (MBOAT7) or LENG4 belongs to membrane-bound O-acyltransferase family. The gene encoding it is localized on human chromosome 19q13.42.
Mouse monoclonal antibody raised against a partial recombinant LENG4.

Immunogen

LENG4 (NP_077274.2, 96 a.a. ~ 192 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TPTPFTNAVQLLLTLKLVSLASEVQDLHLAQRKEMASGFSKGPTLGLLPDVPSLMETLSYSYCYVGIMTGPFFRYRTYLDWLEQPFPGAVPSLRPLL

Biochem/physiol Actions

Membrane bound O-acyltransferase domain containing 7 (MBOAT7) or LENG4 is an integral membrane protein important for reacylation of phospholipids. LENG4 is highly specific for arachidonoyl-coenzyme A and is involved in arachidonate recycling. It interacts with the small subunit of serine palmitoyltransferase a (ssSPTa). This interaction is important for LENG4-dependent incorporation of arachidonic acid into phosphatidylinositol.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent progress on acyl CoA: lysophospholipid acyltransferase research.
Shindou H
Journal of Lipid Research (2009)
Yusuke Hirata et al.
Genes to cells : devoted to molecular & cellular mechanisms, 18(5), 397-409 (2013-03-21)
Lysophosphatidylinositol acyltransferase 1 (LPIAT1), also known as MBOAT7, is a phospholipid acyltransferase that selectively incorporates arachidonic acid (AA) into the sn-2 position of phosphatidylinositol (PI). We previously demonstrated that LPIAT1 regulates AA content in PI and plays a crucial role
Miguel A Gijón et al.
The Journal of biological chemistry, 283(44), 30235-30245 (2008-09-06)
The cycle of deacylation and reacylation of phospholipids plays a critical role in regulating availability of arachidonic acid for eicosanoid production. The major yeast lysophospholipid acyltransferase, Ale1p, is related to mammalian membrane-bound O-acyltransferase (MBOAT) proteins. We expressed four human MBOATs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service