Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402964

Sigma-Aldrich

Monoclonal Anti-ZNF23 antibody produced in mouse

clone 2D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

KOX16, MGC57283, ZNF359, ZNF612, Zfp612

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF23(7571)

General description

The gene ZNF23 (zinc finger protein 23) is mapped to human chromosome 16q22, a region associated with frequent alterations in acute myeloid leukemia. It belongs to the family of KRAB–ZFPs (Krupple-associated box-containing zinc-finger proteins), a large family of transcription factors. These proteins contain a KRAB domain at the N-terminal. The C-terminal consists of 16 tandem C2H2 class zinc fingers. The protein is ubiquitously expressed and the levels are greatly decreased or lost in cases of human cancer. The gene spans a length of 3271 nucleotides that encodes a protein of 643 amino acids.

Immunogen

ZNF23 (NP_666016.1, 41 a.a. ~ 139 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GLQGLQTDIQTDNDLTKEMYEGKENVSFELQRDFSQETDFSEASLLEKQQEVHSAGNIKKEKSNTIDGTVKDETSPVEECFFSQSSNSYQCHTITGEQP

Biochem/physiol Actions

The gene ZNF23 (zinc finger protein 23) encodes a nuclear protein that is involved in the inhibition of cell cycle progression. Ectopic expression of this protein is associated with increased p27kip-1 expression, growth inhibition and cell cycle arrest in G1 phase. The gene functions as a tumor suppressor, as its down regulation has been associated with carcinogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of ZNF23, a KRAB-containing protein that is downregulated in human cancers and inhibits cell cycle progression.
Huang C
Experimental Cell Research, 313, 254-263 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service