Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402393

Sigma-Aldrich

Monoclonal Anti-WNT5A antibody produced in mouse

clone 3A4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

hWNT5A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WNT5A(7474)

General description

Wnt family member 5A (WNT5A) is a secreted, proinflammatory protein. It is part of the noncanonical WNT family. The gene encoding this protein is localized on human chromosome 3p14.3.

Immunogen

WNT5A (AAH64694, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ERERIHAKGSYESARILMNLHNNEAGRRTVYNLADVACKCHGVSGSCSLKTCWLQLADFRKVGDALKEKYDSAAAMRLNSRGKLVQVNSRFNSPTTQDLV

Biochem/physiol Actions

Wnt family member 5A (WNT5A) has a role in metabolic dysfunction in obesity and negatively modulates angiogenesis in adipose tissue. The protein regulates cell motility and polarity by activating β-catenin–independent pathways. It has been shown to be overexpressed in gliomas.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptional mechanisms of WNT5A based on NF-?B, Hedgehog, TGF?, and Notch signaling cascades
Masuko Katoh
International Journal of Molecular Medicine, 763-769 (2009)
Wnt5a Drives an Invasive Phenotype in Human Glioblastoma Stem-like Cells.
Binda E, et.al
Cancer Research, 77(4), 996-1007 (2017)
WNT5A regulates adipose tissue angiogenesis via antiangiogenic VEGF-A165b in obese humans.
Karki S
American Journal of Physiology. Heart and Circulatory Physiology, 313(1) (2017)
Elena Binda et al.
Cancer research, 77(4), 996-1007 (2016-12-25)
Brain invasion by glioblastoma determines prognosis, recurrence, and lethality in patients, but no master factor coordinating the invasive properties of glioblastoma has been identified. Here we report evidence favoring such a role for the noncanonical WNT family member Wnt5a. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service