Skip to Content
Merck
All Photos(1)

Key Documents

SAB1402314

Sigma-Aldrich

Monoclonal Anti-PMP22 antibody produced in mouse

clone 3G10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CMT1A, CMT1E, DSS, GAS-3, HMSNIA, HNPP, MGC20769, Sp110

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.9 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PMP22(5376)

General description

This gene encodes an integral membrane protein that is a major component of myelin in the peripheral nervous system. Various mutations of this gene are causes of Charcot-Marie-Tooth disease Type IA, Dejerine-Sottas syndrome, and hereditary neuropathy with liability to pressure palsies. Alternative splicing of this gene results in three transcript variants that encode the same protein. (provided by RefSeq)

Immunogen

PMP22 (AAH19040, 25 a.a. ~ 114 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VSQWIVGNGHATDLWQNCSTSSSGNVHHCFSSSPNEWLQSVQATMILSIIFSILSLFLFFCQLFTLTKGGRFYITGIFQILAGLCVMSAA

Biochem/physiol Actions

Peripheral myelin protein 22 (PMP22) plays a vital role in myelination during peripheral nerve development. It also has a role in cell-cell interactions, cell proliferation, maintenance of axons and the determination of myelin thickness and stability. Aberrations or mutations in the PMP22 gene lead to heritable demyelinating peripheral neuropathies, such as Charcot-Marie-tooth disease type IA (CMT1A) and Dejerine-Sottas syndrome. Overexpression of PMP22 might contribute to the development of chronic myeloid leukemia (CML).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypermyelination and demyelinating peripheral neuropathy in Pmp22-deficient mice.
Adlkofer K
Nature Genetics, 11(3), 274-280 (1995)
Many facets of the peripheral myelin protein PMP22 in myelination and disease.
Naef R and Suter U
Microscopy Research and Technique, 41(5), 359-371 (1998)
Knockdown of Peripheral Myelin Protein 22 Inhibits the Progression of Chronic Myeloid Leukemia.
Liu H
Oncology Research, 259(65), 259-265 (2014)
Dejerine-Sottas syndrome associated with point mutation in the peripheral myelin protein 22 (PMP22) gene.
Roa BB
Nature Genetics, 5(3), 269-273 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service