Skip to Content
Merck
All Photos(4)

Key Documents

SAB1402289

Sigma-Aldrich

Monoclonal Anti-NFIB antibody produced in mouse

clone 2D6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HMGIC/NFIB, NFI-RED, NFIB2, NFIB3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~72.31 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFIB(4781)

Immunogen

NFIB (AAH01283, 1 a.a. ~ 420 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MMYSPICLTQDEFHPFIEALLPHVRAIAYTWFNLQARKRKYFKKHEKRMSKDEERAVKDELLSEKPEIKQKWASRLLAKLRKDIRQEYREDFVLTVTGKKHPCCVLSNPDQKGKIRRIDCLRQADKVWRLDLVMVILFKGIPLESTDGERLMKSPHCTNPALCVQPHHITVSVKELDLFLAYYVQEQDSGQSGSPSHNDPAKNPPGYLEDSFVKSGVFNVSELVRVSRTPITQGTGVNFPIGEIPSQPYYHDMNSGVNLQRSLSSPPSSKRPKTISIDENMEPSPTGDFYPSPSSPAAGSRTWHERDQDMSSPTTMKKPEKPLFSSASPQDSSPRLSTFPQHHHPGIPGVAHSVISTRTPPPPSPLPFPTQAILPPAPSSYFSHPTIRYPPHLNPQDTLKNYVPSYDPSSPQTSQSWYLG

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service