Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401732

Sigma-Aldrich

Anti-AHI1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AHI-1, DKFZp686J1653, FLJ14023, FLJ20069, JBTS3, ORF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHI1(54806)

General description

This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified. (provided by RefSeq)

Immunogen

AHI1 (AAH94800.1, 1 a.a. ~ 609 a.a) full-length human protein.

Sequence
MPTAESEAKVKTKVRFEELLKTHSDLMREKKKLKKKLVRSEENISPDTIRSNLHYMKETTSDDPDTIRSNLPHIKETTSDDVSAANTNNLKKSTRVTKNKLRNTQLATENPNGDASVEEDKQGKPNKKVIKTVPQLTTQDLKPETPENKVDSTHQKTHTKPQPGVDHQKSEKANEGREETDLEEDEELMQAYQCHVTEEMAKEIKRKIRKKLKEQLTYFPSDTLFHDDKLSSEKRKKKKEVPVFSKAETSTLTISGDTVEGEQKKESSVRSVSSDSHQDDEISSMEQSTEDSMQDDTKPKPKKTKKKTKAVADNNEDVDGDGVHEITSRDSPVYPKCLLDDDLVLGVYIHRTDRLKSDFMISHPMVKIHVVDEHTGQYVKKDDSGRPVSSYYEKENVDYILPIMTQPYDFKQLKSRLPEWEEQIVFNENFPYLLRGSDESPKVILFFEILDFLSVDEIKNNSEVQNQECGFRKIAWAFLKLLGANGNANINSKLRLQLYYPPTKPRSPLSVVEAFEWWSKCPRNHYPSTLYVTVRGLKVPDCIKPSYRSMMALQEEKGKPVHCERHHESSSVDTEPGLEESKEVIKWKRLPGQENEDVFVLISPTMEEY

Biochem/physiol Actions

The AHI1 (Abelson helper integration site 1) protein is responsible for the cerebellar and cortical structure growth during embryonic development. Mutation in AHI1 (Abelson helper integration site 1) might result in schizophrenia, a mental disorder that is highly heritable. AHI1 variants cause oculomotor apraxia, leading to Joubert syndrome-related disorders. The encoded protein is associated with canonical Wnt signaling.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of 2 Potentially Relevant Gene Mutations Involved in Strabismus Using Whole-Exome Sequencing.
Min X
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 23, 1719-1724 (2017)
A cis-eQTL in AHI1 confers risk to schizophrenia in European populations.
Ren Z, et.al.
Neuroscience Letters, 632, 130-135 (2016)
Zhimin Ren et al.
Neuroscience letters, 632, 130-135 (2016-09-03)
Schizophrenia is a devastating mental disorder, with heritability as high as 80%. Although genome-wide association studies have identified multiple promising risk variants of schizophrenia, they could only explain a small portion of the disease heritability, and other variants with low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service