Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401644

Sigma-Aldrich

Anti-SESN1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC138241, MGC142129, PA26, RP11-787I22.1, SEST1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SESN1(27244)

Immunogen

SESN1 (NP_055269.1, 1 a.a. ~ 551 a.a) full-length human protein.

Sequence
MAEGENEVRWDGLCSRDSTTRETALENIRQTILRKTEYLRSVKETPHRPSDGLSNTESSDGLNKLLAHLLMLSKRCPFKDVREKSEFILKSIQELGIRIPRPLGQGPSRFIPEKEILQVGSEDAQMHALFADSFAALGRLDNITLVMVFHPQYLESFLKTQHYLLQMDGPLPLHYRHYIGIMAAARHQCSYLVNLHVNDFLHVGGDPKWLNGLENAPQKLQNLGELNKVLAHRPWLITKEHIEGLLKAEEHSWSLAELVHAVVLLTHYHSLASFTFGCGISPEIHCDGGHTFRPPSVSNYCICDITNGNHSVDEMPVNSAENVSVSDSFFEVEALMEKMRQLQECRDEEEASQEEMASRFEIEKRESMFVFSSDDEEVTPARAVSRHFEDTSYGYKDFSRHGMHVPTFRVQDYCWEDHGYSLVNRLYPDVGQLIDEKFHIAYNLTYNTMAMHKDVDTSMLRRAIWNYIHCMFGIRYDDYDYGEINQLLDRSFKVYIKTVVCTPEKVTKRMYDSFWRQFKHSEKVHVNLLLIEARMQAELLYALRAITRYMT

Biochem/physiol Actions

Sesn1 (sestrin 1) is known to be a protein involved in sensing stress. It promotes the AMPK (5′ AMP-activated protein kinase) activation. Sesn1 inhibits mTORC1 (mammalian target of rapamycin complex 1) signaling, thereby contributing to anti-ageing mechanism. Sestrins protein family possesses oxidoreductase activity. Sestrins are believed to promote antioxidative responses to stress, as they are transcripted by p53 (tumor protein) and FoxO (Forkhead box). Thus, they can serve as a potential therapeutic target.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapamycin attenuates endothelial apoptosis induced by low shear stress via mTOR and sestrin1 related redox regulation.
Zhang J
Mediators of Inflammation, 2014:769608, 1-9 (2014)
A sestrin-dependent Erk-Jnk-p38 MAPK activation complex inhibits immunity during aging.
Lanna A
Nature Immunology, 18(3), 354-363 (2017)
Regeneration of peroxiredoxins by p53-regulated sestrins, homologs of bacterial AhpD.
Budanov AV
Science, 304(5670), 596-600 (2004)
Irena Horwacik et al.
Acta biochimica Polonica, 62(3), 423-433 (2015-08-19)
Neuroblastoma is the most common extra-cranial solid tumor of childhood and it is characterized by the presence of a glycosphingolipid, GD2 ganglioside. Monoclonal antibodies targeting the antigen are currently tested in clinical trials. Additionally, several research groups reported results revealing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service