Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401588

Sigma-Aldrich

Anti-ABCB9 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

EST122234, KIAA1520, TAPL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCB9(23457)

General description

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. The function of this half-transporter has not yet been determined; however, this protein may play a role in lysosomes. Alternative splicing of this gene results in distinct isoforms which are likely to have different substrate specifications. (provided by RefSeq)

Immunogen

ABCB9 (AAH17348.1, 1 a.a. ~ 596 a.a) full-length human protein.

Sequence
MRLWKAVVVTLAFMSVDICVTTAIYVFSHLDRSLLEDIRHFNIFDSVLDLWAACLYRSCLLLGATIGVAKNSALGPRRLRASWLVITLVCLFVGIYAMVKLLLFSEVRRPIRDPWFWALFVWTYISLGASFLLWWLLSTVRPGTQALEPGAATEAEGFPGSGRPPPEQASGATLQKLLSYTKPDVAFLVAASFFLIVAALGETFLPYYTGRAIDGIVIQKSMDQFSTAVVIVCLLAIGSSFAAGIRGGIFTLIFARLNIRLRNCLFRSLVSQETSFFDENRTGDLISRLTSDTTMVSDLVSQNINVFLRNTVKVTGVVVFMFSLSWQLSLVTFMGFPIIMMVSNIYGKYYKRLSKEVQNALARASNTAEETISAMKTVRSFANEEEEAEVYLRKLQQVYKLNRKEAAAYMYYVWGSGLTLLVVQVSILYYGGHLVISGQMTSGNLIAFIIYEFVLGDCMESVGSVYSGLMQGVGAAEKVFEFIDRQPTMVHDGSLAPDHLEGRVDFENVTFTYRTRPHTQVLQNVSFSLSPGKVTALVGPSGSGKSSCVNILENFYPLEGGRVLLDGKPISAYDHKYLHRVVCARAWATLLRPFCI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service