Skip to Content
Merck
All Photos(5)

Key Documents

SAB1401559

Sigma-Aldrich

Monoclonal Anti-ATE1 antibody produced in mouse

clone 2B6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC26724

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATE1(11101)

General description

This gene encodes an arginyltransferase, an enzyme that is involved in posttranslational conjugation of arginine to N-terminal aspartate or glutamate residues. Conjugation of arginine to the N-terminal aspartate or glutamate targets proteins for ubiquitin-dependent degradation. Alternative splicing results in two transcript variants encoding distinct isoforms. (provided by RefSeq)

Immunogen

ATE1 (NP_001001976, 1 a.a. ~ 87 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAFWAGGSPSVVDYFPSEDFYRCGYCKNESGSRSNGMWAHSMTVQDYQDLIDRGWRRSGKYVYKPVMNQTCCPQYTIRCRPLQFQPS

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service