Skip to Content
Merck
All Photos(1)

Documents

SAB1401121

Sigma-Aldrich

Monoclonal Anti-LPAR1 antibody produced in mouse

clone 2E2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LPAR1(1902)

Related Categories

General description

The integral membrane protein encoded by this gene is a lysophosphatidic acid (LPA) receptor from a group known as EDG receptors. These receptors are members of the G protein-coupled receptor superfamily. Utilized by LPA for cell signaling, EDG receptors mediate diverse biologic functions, including proliferation, platelet aggregation, smooth muscle contraction, inhibition of neuroblastoma cell differentiation, chemotaxis, and tumor cell invasion. Two transcript variants encoding the same protein have been identified for this gene (provided by RefSeq)

Immunogen

LPAR1 (AAH30615, 1 a.a. ~ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAAISTSIPVISQPQFTAMNEPQCFYNESIAFFYNRSGKHLATEWNTVSKLVMGLGITVCIFIMLANLLVMVAIYVNRRFHFPIYYLMANLAAADFFAGLAYFYLMFNTGPNTRRLTVSTWLLRQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bumjun Kim et al.
Molecular pharmaceutics, 16(5), 1813-1826 (2019-03-19)
The plasticity of cancer epigenetics makes them plausible candidates for therapeutic intervention. We took advantage of elevated expression of lysophosphatidic acid receptor 1 (LPAR1) in triple negative breast cancer (TNBC) tissues to target decitabine (DAC) and panobinostat (PAN) to breast
Wentao Zhong et al.
Experimental cell research, 379(1), 65-72 (2019-03-23)
Olfactory ensheathing cells (OECs) are ideal candidates for cell-based therapies aimed at repairing spinal cord injury (SCI). Accurate targeting of OECs to the lesion site is critical to reconstructing the impaired nervous system. However, the key factors guiding the homing
Jong Han Lee et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(6), 1332-1340 (2019-02-15)
Diabetic nephropathy (DN) is one of the major long-term complications of diabetes. Lysophosphatidic acid (LPA) signaling has been implicated in renal fibrosis. In our previous study, we found that the LPA receptor 1/3 (LPAR1/3) antagonist, ki16425, protected against DN in
Vijay Walia et al.
Developmental cell, 50(2), 229-246 (2019-06-18)
Serum starvation stimulates cilia growth in cultured cells, yet serum factors associated with ciliogenesis are unknown. Previously, we showed that starvation induces rapid Rab11-dependent vesicular trafficking of Rabin8, a Rab8 guanine-nucleotide exchange factor (GEF), to the mother centriole, leading to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service