Skip to Content
Merck
All Photos(3)

Key Documents

SAB1401069

Sigma-Aldrich

Anti-CENPB antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CENPB(1059)

General description

Centromere protein B (CENPB) is a dimeric protein, localized to the centromeric chromatin. It possesses a DNA-binding activity. The protein has a DNA-binding domain at the amino-terminal region and a dimerization domain at the carboxy-terminal region. The subunits of CENPB have a molecular weight of 80kDa. The gene encoding CENPB is localized on human chromosome 20.

Immunogen

CENPB (AAH53847.1, 1 a.a. ~ 599 a.a) full-length human protein.

Sequence
MGPKRRQLTFREKSRIIQEVEENPDLRKGEIARRFNIPPSTLSTILKNKRAILASERKYGVASTCRKTNKLSPYDKLEGLLIAWFQQIRAAGLPVKGIILKEKALRIAEELGMDDFTASNGWLDRFRRRHGVVSCSGVARARARNAAPRTPAAPASPAAVPSEGSGGSTTGWRAREEQPPSVAEGYASQDVFSATETSLWYDFLPDQAAGLCGGDGRPRQATQRLSVLLCANADGSEKLPPLVAGKSAKPRAGQAGLPCDYTANSKGGVTTQALAKYLKALDTRMAAESRRVLLLAGRLAAQSLDTSGLRHVQLAFFPPGTVHPLERGVVQQVKGHYRQAMLLKAMAALEGQDPSGLQLGLTEALHFVAAAWQAVEPSDIAACFREAGFGGGPNATITTSLKSEGEEEEEEEEEEEEEEGEGEEEEEEGEEEEEEGGEGEELGEEEEVEEEGDVDSDEEEEEDEESSSEGLEAEDWAQGVVEAGGSFGAYGAQEEAQCPTLHFLEGGEDSDSDSEEEDDEEEDDEDEDDDDDEEDGDEVPVPSFGEAMAYFAMVKRYLTSFPIDDRVQSHILHLEHDLVHVTRKNHARQAGVRGLGHQS

Biochem/physiol Actions

Centromere protein B (CENPB) has a role in assembly of the components of the centromere. It binds to the 17-base pair sequence called the CENP-B box on the DNA.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping of the human centromere protein B gene (CENPB) to chromosome 20p13 by fluorescence in situ hybridization
Naohiko S, et al.
Genomics, 24(1). (1994)
Mutational spectrum of CENP-B box and a-satellite DNA on chromosome 21 in Down syndrome children.
Chen Q, et al.
Molecular Medicine Reports, 15(4), 2313-2317 (2017)
HTLV-1 bZIP factor suppresses the centromere protein B (CENP-B)-mediated trimethylation of histone H3K9 through the abrogation of DNA-binding ability of CENP-B.
Mukai R, et al.
The Journal of General Virology, 96(Pt 1), 159-164 (2015)
Human centromere protein B induces translational positioning of nucleosomes on alpha-satellite sequences.
Tanaka Y, et al.
The Journal of Biological Chemistry, 280(50), 41609-41618 (2005)
Stable complex formation of CENP-B with the CENP-A nucleosome.
Fujita R, et al.
Nucleic Acids Research, 43(10), 4909-4022 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service