Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400507

Sigma-Aldrich

Anti-APEX2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-APE2, Anti-APEXL2, Anti-XTH2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APEX2(27301)

General description

Apurinic/apyrimidinic (AP) sites occur frequently in DNA molecules by spontaneous hydrolysis, by DNA damaging agents or by DNA glycosylases that remove specific abnormal bases. AP sites are pre-mutagenic lesions that can prevent normal DNA replication so the cell contains systems to identify and repair such sites. Class II AP endonucleases cleave the phosphodiester backbone 5′ to the AP site. This gene encodes a protein shown to have a weak class II AP endonuclease activity. Most of the encoded protein is located in the nucleus but some is also present in mitochondria. This protein may play an important role in both nuclear and mitochondrial base excision repair (BER). (provided by RefSeq)

Immunogen

APEX2 (NP_055296.2, 1 a.a. ~ 518 a.a) full-length human protein.

Sequence
MLRVVSWNINGIRRPLQGVANQEPSNCAAVAVGRILDELDADIVCLQETKVTRDALTEPLAIVEGYNSYFSFSRNRSGYSGVATFCKDNATPVAAEEGLSGLFATQNGDVGCYGNMDEFTQEELRALDSEGRALLTQHKIRTWEGKEKTLTLINVYCPHADPGRPERLVFKMRFYRLLQIRAEALLAAGSHVIILGDLNTAHRPIDHWDAVNLECFEEDPGRKWMDSLLSNLGCQSASHVGPFIDSYRCFQPKQEGAFTCWSAVTGARHLNYGSRLDYVLGDRTLVIDTFQASFLLPEVMGSDHCPVGAVLSVSSVPAKQCPPLCTRFLPEFAGTQLKILRFLVPLEQSPVLEQSTLQHNNQTRVQTCQNKAQVRSTRPQPSQVGSSRGQKNLKSYFQPSPSCPQASPDIELPSLPLMSALMTPKTPEEKAVAKVVKGQAKTSEAKDEKELRTSFWKSVLAGPLRTPLCGGHREPCVMRTVKKPGPNLGRRFYMCARPRGPPTDPSSRCNFFLWSRPS

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service