Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400377

Sigma-Aldrich

Anti-RBM6 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-3G2, Anti-DEF-3, Anti-DEF3, Anti-NY-LU-12, Anti-g16

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€369.00

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€369.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBM6(10180)

General description

RNA binding motif protein 6 (RBM6) is expressed in peripheral blood leukocytes, thymus and lymph nodes. The gene encoding it is localized on human chromosome 3p21.3 and has 21 exons.

Immunogen

RBM6 (AAH46643.1, 1 a.a. ~ 601 a.a) full-length human protein.

Sequence
MIQDKEVTLEYVSSLDFWYCKRCKANIGGHRSSCSFCKNPREVTEAKQELITYPQPQKTSIPAPLEKQPNQPLRPADKEPEPRKREEGQESRLGHQKREAERYLPPSRREGPTFRRDRERESWSGETRQDGESKTIMLKRIYRSTPPEVIVEVLEPYVRLTTANVRIIKNRTGPMGHTYGFIDLDSHAEALRVVKILQNLDPPFSIDGKMVAVNLATGKRRNDSGDHSDHMHYYQGKKYFRDRRGGGRNSDWSSDTNRQGQQSSSDCYIYDSATGYYYDPLAGTYYDPNTQQEVYVPQDPGLPEEEEIKEKKPTSQGKSSSKKEMSKRDGKEKKDRGVTRFQENASEGKAPAEDVFKKPLPPTVKKEESPPPPKVVNPLIGLLGEYGGDSDYEEEEEEEQTPPPQPRTAQPQKREEQTKKENEEDKLTDWNKLACLLCRRQFPNKEVLIKHQQLSDLHKQNLEIHRKIKQSEQELAYLERREREGKFKGRGNDRREKLQSFDSPERKRIKYSRETDSDRKLVDKEDIDTSSKGGCVQQATGWRKGTGLGYGHPGLASSEEAEGRMRGPSVGASGRTSKRQSNETYRDAVRRVMFARYKELD

Biochem/physiol Actions

RNA binding motif protein 6 (RBM6) associates with RNA. It has been linked to lung cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elias G Bechara et al.
Molecular cell, 52(5), 720-733 (2013-12-18)
RBM5, a regulator of alternative splicing of apoptotic genes, and its highly homologous RBM6 and RBM10 are RNA-binding proteins frequently deleted or mutated in lung cancer. We report that RBM5/6 and RBM10 antagonistically regulate the proliferative capacity of cancer cells
Ting-lei Gu et al.
Blood, 110(1), 323-333 (2007-03-16)
Activated tyrosine kinases have been frequently implicated in the pathogenesis of cancer, including acute myeloid leukemia (AML), and are validated targets for therapeutic intervention with small-molecule kinase inhibitors. To identify novel activated tyrosine kinases in AML, we used a discovery
Ke Wang et al.
BMC genomics, 8, 348-348 (2007-10-03)
Transcription-induced chimerism, a mechanism involving the transcription and intergenic splicing of two consecutive genes, has recently been estimated to account for approximately 5% of the human transcriptome. Despite this prevalence, the regulation and function of these fused transcripts remains largely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service