Skip to Content
Merck
All Photos(3)

Documents

SAB1400337

Sigma-Aldrich

Anti-CBFA2T2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DKFZp313F2116, Anti-EHT, Anti-MTGR1, Anti-ZMYND3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBFA2T2(9139)

General description

In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5′-region of the RUNX1 (AML1) gene fused to the 3′-region of the CBFA2T1 (MTG8) gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene binds to the AML1-MTG8 complex and may be important in promoting leukemogenesis. Several transcript variants are thought to exist for this gene, but the full-length natures of only three have been described. (provided by RefSeq)

Immunogen

CBFA2T2 (NP_001034798.1, 1 a.a. ~ 575 a.a) full-length human protein.

Sequence
MPGSPVEVKIQSRSSPPTMPPLPPINPGGPRPVSFTPTALSNGINHSPPTLNGAPSPPQRFSNGPASSTSSALTNQQLPATCGARQLSKLKRFLTTLQQFGNDISPEIGEKVRTLVLALVNSTVTIEEFHCKLQEATNFPLRPFVIPFLKANLPLLQRELLHCARAAKQTPSQYLAQHEHLLLNTSIASPADSSELLMEVHGNGKRPSPERREENSFDRDTIAPEPPAKRVCTISPAPRHSPALTVPLMNPGGQFHPTPPPLQHYTLEDIATSHLYREPNKMLEHREVRDRHHSLGLNGGYQDELVDHRLTEREWADEWKHLDHALNCIMEMVEKTRRSMAVLRRCQESDREELNYWKRRYNENTELRKTGTELVSRQHSPGSADSLSNDSQREFNSRPGTGYVPVEFWKKTEEAVNKVKIQAMSEVQKAVAEAEQKAFEVIATERARMEQTIADVKRQAAEDAFLVINEQEESTENCWNCGRKASETCSGCNIARYCGSFCQHKDWERHHRLCGQNLHGQSPHGQGRPLLPVGRGSSARSADCSVPSPALDKTSATTSRSSTPASVTAIDTNGL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service