Skip to Content
Merck
All Photos(4)

Key Documents

SAB1400203

Sigma-Aldrich

Anti-PFKP antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FLJ40226, Anti-PFK-C, Anti-PFKF

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PFKP(5214)

Related Categories

General description

The PFKP gene encodes the platelet isoform of phosphofructokinase (PFK) (ATP:D-fructose-6-phosphate-1-phosphotransferase, EC 2.7.1.11). PFK catalyzes the irreversible conversion of fructose-6-phosphate to fructose-1,6-bisphosphate and is a key regulatory enzyme in glycolysis. The PFKP gene, which maps to chromosome 10p, is also expressed in fibroblasts. See also the muscle (PFKM; MIM 610681) and liver (PFKL; MIM 171860) isoforms of phosphofructokinase, which map to chromosomes 12q13 and 21q22, respectively. Vora (1981) [PubMed 6451249] determined that full tetrameric phophofructokinase enzyme expressed in platelets can be composed of subunits P4, P3L, and P2L2.[supplied by OMIM

Immunogen

PFKP (NP_002618.1, 1 a.a. ~ 784 a.a) full-length human protein.

Sequence
MDADDSRAPKGSLRKFLEHLSGAGKAIGVLTSGGDAQGMNAAVRAVVRMGIYVGAKVYFIYEGYQGMVDGGSNIAEADWESVSSILQVGGTIIGSARCQAFRTREGRLKAACNLLQRGITNLCVIGGDGSLTGANLFRKEWSGLLEELARNGQIDKEAVQKYAYLNVVGMVGSIDNDFCGTDMTIGTDSALHRIIEVVDAIMTTAQSHQRTFVLEVMGRHCGYLALVSALACGADWVFLPESPPEEGWEEQMCVKLSENRARKKRLNIIIVAEGAIDTQNKPITSEKIKELVVTQLGYDTRVTILGHVQRGGTPSAFDRILASRMGVEAVIALLEATPDTPACVVSLNGNHAVRLPLMECVQMTQDVQKAMDERRFQDAVRLRGRSFAGNLNTYKRLAIKLPDDQIPKTNCNVAVINVGAPAAGMNAAVRSAVRVGIADGHRMLAIYDGFDGFAKGQIKEIGWTDVGGWTGQGGSILGTKRVLPGKYLEEIATQMRTHSINALLIIGGFEAYLGLLELSAAREKHEEFCVPMVMVPATVSNNVPGSDFSIGADTALNTITDTCDRIKQSASGTKRRVFIIETMGGYCGYLANMGGLAAGADAAYIFEEPFDIRDLQSNVEHLTEKMKTTIQRGLVLRNESCSENYTTDFIYQLYSEEGKGVFDCRKNVLGHMQQGGAPSPFDRNFGTKISARAMEWITAKLKEARGRGKKFTTDDSICVLGISKRNVIFQPVAELKKQTDFEHRIPKEQWWLKLRPLMKILAKYKASYDVSDSGQLEHVQPWSV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service