Skip to Content
Merck
All Photos(2)

Documents

SAB1400013

Sigma-Aldrich

Anti-SLC25A5 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-2F1, Anti-ANT2, Anti-T2, Anti-T3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC25A5(292)

General description

This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein forms a homodimer embedded in the inner mitochondria membrane. Suppressed expression of this gene has been shown to induce apoptosis and inhibit tumor growth. The human genome contains several non-transcribed pseudogenes of this gene. Solute carrier family 25 member 5 (SLC25A5) is a mitochondrial protein, which promotes the exchange of ADP and ATP across the inner mitochondrial membrane. SLC25A5 is located on human chromosome Xq24. It is found mainly in proliferative and undifferentiated cells.

Immunogen

SLC25A5 (AAH56160, 1 a.a. ~ 298 a.a) full-length human protein.

Sequence
MTDAAVSFAKDFLAGGVAAAISKTAVAPIERVKLLLQVQHASKQITADKQYKGIIDCVVRIPKEQGVLSFWRGNLANVIRYFPTQALNFAFKDKYKQIFLGGVDKRTQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKAGAEREFRGLGDCLVKIYKSDGIKGLYQGFNVSVQGIIIYRAAYFGIYDTAKGMLPDPKNTHIVISWMIAQTVTAVAGLTSYPFDTVRRRMMMQSGRKGTDIMYTGTLDCWRKIARDEGGKAFFKGAWSNVLRGMGGAFVLVLYDEIKKYT

Biochem/physiol Actions

Solute carrier family 25 member 5 (SLC25A5) is necessary for cellular energy metabolism. It transports ATP by glycolysis. SLC25A5 is an indicator of carcinogenesis. SLC25A5 is associated with non-syndromic intellectual disability. Silencing of this gene induces apoptosis and inhibits tumor growth.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenine nucleotide translocase 2 is a key mitochondrial protein in cancer metabolism
Chevrollier A, et al.
Biochimica et Biophysica Acta - Bioenergetics, 1807(6) (2011)
Suppression of adenine nucleotide translocase-2 by vector-based siRNA in human breast cancer cells induces apoptosis and inhibits tumor growth in vitro and in vivo
Jang J, et al.
Breast Cancer Research, 10(1), R11-R11 (2008)
ANT2-defective fibroblasts exhibit normal mitochondrial bioenergetics
Prabhu D, et al.
Molecular genetics and metabolism reports, 3 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service