Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306481

Sigma-Aldrich

ANTI-CD45(C-TERMINAL) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

CD45, Leukocyte common antigen, PTPRC, Receptor-type tyrosine-protein phosphatase C, T200

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€388.00

€388.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1180


Select a Size

Change View
400 μL
€388.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€388.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1180

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

147254 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)

Related Categories

General description

Protein tyrosine phosphatase, receptor type C (PTPRC) or leucocyte common antigen CD45 is expressed on T-cells and is found in multiple isoforms. The extracellular domain of the protein is predicted to have an amino-terminal mucin-like segment, a cysteine-rich domain and three fibronectin type 3 domains. The gene encoding this protein is localized on human chromosome 1q31.3.

Application

ANTI-CD45(C-TERMINAL) antibody produced in rabbit has been used in Western blotting.

Biochem/physiol Actions

Protein tyrosine phosphatase, receptor type C (PTPRC) or leucocyte common antigen CD45 acts as a protein tyrosine phosphatase in leucocyte signaling. It has a role in flow cytometric diagnostics in leukemia. By dephosphorylation of Src kinases, the protein acts as a regulator of antigen receptor signaling in T and B cells. As a negative regulator of janus kinases, CD45 inhibits cytokine receptor signaling.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High CD45 surface expression determines relapse risk in children with precursor B-cell and T-cell acute lymphoblastic leukemia treated according to the ALL-BFM 2000 protocol
Gunnar Cario, et al.
Haematologica (2014)
Initiation of T cell signaling by CD45 segregation at `close-contacts?
Veronica T, et al.
Nature Immunology (2016)
CD45RO+ Memory T Lymphocytes ? a Candidate Marker for TNM-Immunoscore in Squamous Non?Small Cell Lung Cancer1
Erna-Elise Paulsen, et al.
Neoplasia (2015)
Clinicoradiological characteristics, management and prognosis of primary myeloid sarcoma of the central nervous system: A report of four cases
Bao Yang, et al.
Oncology Letters (2017)
Replication of association of the PTPRC gene with response to anti?tumor necrosis factor therapy in a large UK cohort*
Darren Plant, et al.
Arthritis & Rheumatology (Hoboken, N.J.) (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service