Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305652

Sigma-Aldrich

ANTI-CD11B(N-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CD11 antigen-like family member B, CD11B, CR3A, ITGAM, Integrin alpha-M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

127179 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGAM(3684)

General description

Cluster of differentiation 11b (CD11b), also known as integrin subunit α M (ITGAM), is encoded by the gene mapped to human chromosome 16p11.2. It belongs to the integrin α chain family. CD11b is expressed by a variety of myeloid cell types, such as dendritic cells, macrophages, monocytes and neutrophils.

Biochem/physiol Actions

Cluster of differentiation 11b (CD11b) acts as a receptor for complement component C3 degradation product, iC3b. CD11b interacts with β2 chain (ITGB2) to form a leukocyte integrin (MAC-1 or complement receptor 3 (CR3)), which in turn plays a crucial role in adherence of neutrophils and monocytes to stimulated endothelium. Mutation in the gene increases the risk of susceptibility to systemic lupus erythematosus (SLE) and acute inflammatory demyelinating polyradiculoneuropathy (AIDP).

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of Systemic Lupus Erythematosus with C8orf13?BLK and ITGAM?ITGAX
Geoffrey Hom,et al.
New. J. Chem. (2008)
Genome-wide association scan in women with systemic lupus erythematosus identifies susceptibility variants in ITGAM, PXK, KIAA1542 and other loci
Harley JB, et al.
Nature Genetics (2008)
A nonsynonymous functional variant in integrin-alpha(M) (encoded by ITGAM) is associated with systemic lupus erythematosus.
Nath SK, et al.
Nature Genetics (2008)
The pathogenic relevance of aM-integrin in Guillain-Barre syndrome.
Acta Neuropathologica (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service