Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305630

Sigma-Aldrich

ANTI-CLEAVED-CASP3 (ASP175) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Apopain, CASP3, CPP32, Caspase-3, Cysteine protease CPP32

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€457.00

€457.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
400 μL
€457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€457.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31608 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Asp175)

Gene Information

human ... CASP3(836)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Feng et al.
Respiratory research, 22(1), 248-248 (2021-09-20)
Chemoresistance limits the therapeutic effect of cisplatin (DDP) on non-small cell lung cancer (NSCLC). Circular RNAs (circRNAs) function as important regulators in chemoresistance. This study aimed to explore the regulation of circRNA Phosphatidylinositol-4-Phosphate 5-Kinase Type 1 Alpha (circ_PIP5K1A) in DDP
Hao Huang et al.
International journal of molecular medicine, 42(5), 2584-2594 (2018-08-15)
The aim of the current study was to explore the potential of TREK‑TRAAK two‑pore domain potassium (K2P) channels in protecting human retinal pigment epithelium (hRPE) cells against oxidative stress. hRPE cells were obtained from donors, and then cell identification and detection of

Questions

  1. What is the sequence of the immunogen used to produce the SAB1305630-400UL antibody? This is needed to check for similarity with the animal model in use.

    1 answer
    1. The antigen region is located within the 149-175 amino acid range. The primary accession number for this information is P42574, which can be found at the UniProtKB database

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service