Skip to Content
Merck
All Photos(1)

Key Documents

SAB1303013

Sigma-Aldrich

ANTI-IL17RB (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Cytokine receptor CRL4, EVI27, IL-17 receptor B, IL-17 receptor homolog 1, IL-17B receptor, IL-17RB, IL-17Rh1, IL17BR, IL17RB, IL17Rh1, Interleukin-17 receptor B, Interleukin-17B receptor, UNQ2501/PRO19612

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€457.00

€457.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
400 μL
€457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€457.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55885 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:250-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL17RB(55540)

General description

Interleukin 17 receptor B (IL-17RB) belongs to the IL-17 receptor family. IL17RB, the receptor for interleukin 17 B (IL17B) is expressed at a high level in the liver, kidney, and other peripheral organs. It is also expressed in memory and effector T lymphocytes. IL17RB gene is located on human chromosome 3p21.1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Interleukin 17 receptor B (IL-17RB) might be considered in thyroid cancer therapy as a potential therapeutic target. IL17RB exhibits much efficient binding avidity to interleukin 17 E (IL17E). IL-17RB is overexpressed in pancreatic, gastric, and prostate cancers.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfonso Lavorgna et al.
PLoS pathogens, 10(10), e1004418-e1004418 (2014-10-24)
Human T-cell leukemia virus type 1 (HTLV-1) infection is linked to the development of adult T-cell leukemia (ATL) and the neuroinflammatory disease HTLV-1 associated myelopathy/tropical spastic paraparesis (HAM/TSP). The HTLV-1 Tax protein functions as a potent viral oncogene that constitutively
Qingli Bie et al.
Scientific reports, 6, 25447-25447 (2016-05-06)
Inflammation is a critical component involved in tumor progression. Interleukin-17 (IL-17) belongs to a relatively new family of cytokines that has been associated with the progression of cancers. However, the role of IL-17B/IL-17RB (IL-17 receptor B) signaling to stemness of
Lei Ren et al.
Molecular immunology, 90, 126-135 (2017-07-18)
IL-17RB, a member of the IL-17 receptor family that can be activated by IL-17B, has been proved to be involved in inflammatory diseases and cancers. However, the function of IL-17RB in thyroid cancer is still unknown. In this study, IL-17RB
Gary M Hunninghake et al.
BMC pulmonary medicine, 11, 17-17 (2011-04-09)
The relationships between total serum IgE levels and gene expression patterns in peripheral blood CD4+ T cells (in all subjects and within each sex specifically) are not known. Peripheral blood CD4+ T cells from 223 participants from the Childhood Asthma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service