Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300929

Sigma-Aldrich

Anti-PHLPP1 (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-EC 3.1.3.16, Anti-PH domain leucine-rich repeat protein phosphatase, Anti-PH domain leucine-rich repeat-containing protein phosphatase, Pleckstrin homology domain-containing family E protein 1, Suprachiasmatic nucleus circadian oscillatory protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€283.00

€283.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μG
€283.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€283.00


Estimated to ship on29 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHLPP1(23239)

General description

Pleckstrin homology domain leucine-rich repeat protein phosphatase 1 (PHLPP1) is encoded by the gene mapped to human chromosome 18q21.33. The encoded protein belongs to the PHLPP family and is characterized with a PH domain followed by region of leucine-rich repeats (LRR), a PP2C phosphatase domain and a C-terminal PDZ ligand. PHLPP1 has two alternatively spliced transcript variants encoding different isoforms namely PHLPP1α with 1205 amino acids and PHLPP1β with 1717 amino acids. PHLPP1 is expressed in cytosolic, nuclear and membrane fraction of cells.

Immunogen

PHLPP1 (NP_919431, 24-58)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human PHLPP1.

Biochem/physiol Actions

PH domain leucine-rich repeat protein phosphatase 1 (PHLPP1) catalyzes the dephosphorylation of serine/threonine-protein kinases AKT2 and AKT3. The encoded protein might act as a tumor suppressor. Loss of gene expression is associated with the development of colon cancers. Overexpression of the gene has been observed in skeletal muscles of Type 2 diabetic patients. Thus, inhibition of PHLPP1 might enhance Akt2 activity and can be considered as a potent therapeutic target for the type II diabetes. Inhibition PHLPP1 through DNA methylation leads to the development of melanoma.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PHLiPPing the switch on Akt and protein kinase C signaling.
Brognard J and Newton A C
Trends in Endocrinology and Metabolism, 19(6), 223-230 (2008)
Low expression of PHLPP1 in sacral chordoma and its association with poor prognosis.
Chen H, et al.
International Journal of Clinical and Experimental Pathology, 8(11), 14741-14741 (2015)
L Dong et al.
Oncogene, 33(39), 4756-4766 (2013-10-15)
Akt is constitutively activated in up to 70% of human melanomas and has an important role in the pathogenesis of the disease. However, little is known about protein phosphatases that dephosphorylate and thereby inactivate it in melanoma cells. Here we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service