Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300728

Sigma-Aldrich

Anti-Phospho-ANTXR1-pY382 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ANTXR1, Anti-ATR, Anti-Anthrax toxin receptor 1 [Precursor], Anti-TEM8, Anti-Tumor endothelial marker 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€283.00

€283.00


Check Cart for Availability


Select a Size

Change View
100 μG
€283.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€283.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

dot blot: suitable
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr382)

Gene Information

human ... ANTXR1(84168)

General description

ANTXR1 is a type I transmembrane protein and is a tumor-specific endothelial marker that has been implicated in colorectal cancer. This protein has been shown to also be a docking protein or receptor for Bacillus anthracis toxin, the causative agent of the disease, anthrax. The binding of the protective antigen (PA) component, of the tripartite anthrax toxin, to this receptor protein mediates delivery of toxin components to the cytosol of cells. Once inside the cell, the other two components of anthrax toxin, edema factor (EF) and lethal factor (LF) disrupt normal cellular processes.

Immunogen

ANTXR1 (NP_115584, 363-396)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y382 of human ANTXR1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service