Skip to Content
Merck
All Photos(3)

Key Documents

SAB1300686

Sigma-Aldrich

Anti-CYP26A1 (center) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-CYP26A1, Cytochrome P450, family 26, subfamily A, polypeptide 1 (Cytochrome P450, family 26, subfamily A, polypeptide 1, isoform CRA_b)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€320.00

€320.00


Estimated to ship on02 May 2025



Select a Size

Change View
100 μG
€320.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€320.00


Estimated to ship on02 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP26A1(1592)

General description

Cytochrome P450 family 26 subfamily A member 1 (CYP26A1) is encoded by the gene mapped to human chromosome 10q23.33.

The previously assigned protein identifier Q5VXI0 has been merged into O43174. Full details can be found on the UniProt database.

Immunogen

CYP26A1 (NP_000774, 123-157)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human CYP26A1.

Biochem/physiol Actions

Cytochrome P450 family 26 subfamily A member 1 (CYP26A1) is a retinoic acid (RA) degrading enzyme. It plays a vital role in RA metabolism. CYP26A1 has an essential role in the correct anteroposterior patterning and the generation of migratory cranial neural crest cells in the developing mammalian brain. Mutations in the gene is associated with the pathogenesis of optic nerve aplasia (ONA).

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical Basis for Dominant Inheritance, Variable Penetrance, and Maternal Effects in RBP4 Congenital Eye Disease.
Chou CM
Cell, 161, 634-646 (2015)
Nonsyndromic bilateral and unilateral optic nerve aplasia: first familial occurrence and potential implication of CYP26A1 and CYP26C1 genes.
Meire F
Molecular Vision, 17, 2072-2079 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service