Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300505

Sigma-Aldrich

Monoclonal Anti-OIT3(L) antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-LZP, Anti-OIT3, Anti-Oncoprotein-induced transcript 3 protein [Precursor], Liver-specific zona pellucida domain-containing protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€377.00

€377.00


Check Cart for Availability


Select a Size

Change View
100 μG
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€377.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

63CT2.5.3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OIT3(170392)

General description

Liver-specific ZP domain-containing protein (OIT3) is a liver-specific protein involved possibly in hepatocellular function and development. GST-tag OIT3 recombinant protein(large) is used to produce this monoclonal antibody.

Immunogen

OIT3 (NP_689848, GST Recombinant Protein (large))
GST-tag OIT3 recombinant protein(large) is used to produce this monoclonal antibody.

Physical form

Purified monoclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service