Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300392

Sigma-Aldrich

Anti-ALPK1 (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-ALPK1, Anti-LAK, Anti-Lymphocyte α-kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€283.00

€283.00


Estimated to ship on02 May 2025



Select a Size

Change View
100 μG
€283.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€283.00


Estimated to ship on02 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALPK1(80216)

Related Categories

General description

Unlike most eukaryotic kinases, alpha kinases, such as ALPK1, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation. Epithelial cells maintain a polarized structure based on a selective sorting machinery for cargo traveling to the apical or the basolateral membrane domain at the trans-Golgi network exit. Alpha-kinase 1 (ALPK1)is a compnent of raft-carrying apical vesicles, originally identified in vesicles ferrying raft-associated sucrase-isomaltase (SI). It has been proposed that phosphorylation of myosin I by ALPK1 is essential to the apical trafficking of raft-associated SI.

Immunogen

ALPK1 (NP_079420, 1220-1254)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human ALPK1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service