Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300358

Sigma-Aldrich

Anti-MTMR2 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€283.00

€283.00


Check Cart for Availability


Select a Size

Change View
100 μG
€283.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€283.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTMR2(8898)

General description

The MTMR2 gene encodes a member of the myotubularin family, characterized by the presence of a phosphatase domain. Mutations in the myotubularin gene (MTM) cause X-linked myotubular myopathy. Northern blot analysis reveals that the 4-kb MTMR2 mRNA is expressed ubiquitously. The mouse Mtmr2 gene encodes a 643-amino acid protein that shares 97% sequence identity with the human protein. Mouse Mtmr2 dephosphorylates phosphatidylinositol 3-phosphate (PI3P) and phosphatidylinositol 3,5-bisphosphate (PI3,5P2), most effectively at neutral pH. This is distinct from the activity of myotubularin, which only acts on PI3P. Analysis of MTMR2 mutations associated with Charcot-Marie-Tooth disease type 4B (CMT4B1) show markedly diminished phosphatase activity, suggesting that this activity is crucial for the proper functioning of peripheral nerves. MTMR2 mutations may lead to malfunction of neural membrane recycling, membrane trafficking, and endo- and exocytic processes. Loss-of-function mutations in MTMR2 are associated with the CMT4B phenotype. MTMR2 interacts with MTMR5, and mutations in the coiled-coil domain of either MTMR2 or MTMR5 abolish this interaction. Through this interaction, MTMR5 promotes enzymatic activity of MTMR2 and prescribe subcellular localization.

Immunogen

MTMR2 (NP_003903, 7-35)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human MTMR2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service