Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300276

Sigma-Aldrich

Anti-ARL2 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ADP-ribosylation factor-like protein 2, Anti-ARL2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€283.00

€283.00


Check Cart for Availability


Select a Size

Change View
100 μG
€283.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€283.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARL2(402)

General description

ADP-ribosylation factors (ARFs) regulate intracellular vesicular membrane trafficking and stimulate a phospholipase D isoform. ARL (ADP-ribosylation like factor) proteins are very similar in sequence to ARFs. ARFs and ARF-like (ARL) proteins, which share sequence similarity with ARFs, form a subfamily of the Ras-related GTPase superfamily. ARL2 encodes a ubiquitiously expressed 184-amino acid predicted protein that is 76% identical to ARF1 and 40 to 45% identical to the Drosophila ARL proteins. Recombinant ARL2 binds to GTP rapidly but guanine nucleotide exchange does not require phospholipids, which is a characteristic of the ARF proteins.

Immunogen

ARL2 (NP_001658, 1-30)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human ARL2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Bi Cai et al.
Clinical genetics, 96(1), 61-71 (2019-04-05)
Adenosine diphosphate (ADP)-ribosylation factor-like 2 (ARL2) protein participates in a broad range of cellular processes and acts as a mediator for mutant ARL2BP in cilium-associated retinitis pigmentosa and for mutant HRG4 in mitochondria-related photoreceptor degeneration. However, mutant ARL2 has not
Xue-Bi Cai et al.
Clinical genetics, 96(1), 61-71 (2019-04-05)
Adenosine diphosphate (ADP)-ribosylation factor-like 2 (ARL2) protein participates in a broad range of cellular processes and acts as a mediator for mutant ARL2BP in cilium-associated retinitis pigmentosa and for mutant HRG4 in mitochondria-related photoreceptor degeneration. However, mutant ARL2 has not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service