Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300233

Sigma-Aldrich

Anti-NOC2L (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-NIR, Anti-NOC2-like, Anti-NOC2L, Anti-Nucleolar complex protein 2 homolog, Anti-Protein NOC2 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€283.00

€283.00


Check Cart for Availability


Select a Size

Change View
100 μG
€283.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€283.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOC2L(26155)

General description

NOC2L is a novel INHAT (inhibitor of histone acetyltransferase). Upon recruitment by p53, this protein represses transcription of both p53-dependent reporters and endogenous target genes.

Immunogen

NOC2L (NP_056473, 474-508)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the central region of human NOC2L.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service