Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300009

Sigma-Aldrich

Anti-HtrA1 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-L56, Anti-Serine protease HTRA1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€377.00

€377.00


Check Cart for Availability


Select a Size

Change View
100 μG
€377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€377.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTRA1(5654)

Related Categories

General description

HtrA1 a member of the trypsin family of serine proteases. This protein is a secreted enzyme that is proposed to regulate the availability of insulin-like growth factors (IGFs) by cleaving IGF-binding proteins. It has also been suggested to be a regulator of cell growth.

Immunogen

HTRA1 (NP_002766, 120-157)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human HtrA1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhu Hou et al.
Arthritis and rheumatism, 65(11), 2835-2846 (2013-08-29)
The protease HTRA-1 is closely associated with rheumatoid arthritis (RA). The molecular mechanisms that control HTRA-1 expression are currently unknown. This study was undertaken to determine the regulatory role of Toll-like receptors (TLRs) on HTRA-1 expression in mice with collagen-induced
Yuzhu Hou et al.
Journal of immunology (Baltimore, Md. : 1950), 193(1), 130-138 (2014-06-08)
The high temperature requirement A1 (HTRA1) is a potent protease involved in many diseases, including rheumatoid arthritis (RA). However, the regulatory mechanisms that control HTRA1 expression need to be determined. In this study, we demonstrated that IFN-γ significantly inhibited the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service