Skip to Content
Merck
All Photos(1)

Key Documents

S9757

Sigma-Aldrich

Sulfabenzamide

Synonym(s):

N-(4-Aminobenzenesulfonyl)benzamide, N-Sulfanilylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2O3S
CAS Number:
Molecular Weight:
276.31
Beilstein:
2139003
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

antibiotic activity spectrum

Gram-negative bacteria

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)NC(=O)c2ccccc2

InChI

1S/C13H12N2O3S/c14-11-6-8-12(9-7-11)19(17,18)15-13(16)10-4-2-1-3-5-10/h1-9H,14H2,(H,15,16)

InChI key

PBCZLFBEBARBBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Physico-chemical comparison of two sulfamides: sulfaproxyline and sulfabenzamide].
L Maury et al.
Pharmaceutica acta Helvetiae, 62(4), 116-120 (1987-01-01)
Nino G Todua et al.
Rapid communications in mass spectrometry : RCM, 25(6), 750-754 (2011-02-22)
Mono-, di- and trialkyl derivatives of 'sulfabenzamide' (N-4-aminophenylsulfonylbenzamide) have been prepared and their electron ionization (EI) mass spectra examined. It is found that the fragmentation of N-alkylsulfabenzamides (alkyl = CH(3) to n-C(5)H(11)) proceeds via a very specific rearrangement process. The proposed mechanism
Ahmad G Nozad et al.
Biophysical chemistry, 139(2-3), 116-122 (2008-11-26)
A systematic computational study was carried out to characterize the hydrogen bond, HB, interactions of sulfabenzamide crystal structure by DFT calculations of electric field gradient, EFG, tensors at the sites of 14N, 17O, and 2H nuclei. The computations were performed
B M Jones et al.
Antimicrobial agents and chemotherapy, 21(6), 870-872 (1982-06-01)
Recent reports suggest that anaerobic Bacteroides organisms are frequently found with Gardnerella vaginalis in nonspecific vaginitis. Specimens taken from 96 women with vaginal discharge were tested simultaneously for these organisms. G. vaginalis was found in 73% of the specimens, Bacteroides
C A Spinks et al.
Bioconjugate chemistry, 10(4), 583-588 (1999-07-20)
Molecular modeling of hapten structure was used to predict and influence, through appropriate synthetic work, the outcome of an immunization program. Examination of the structures of sulfonamide antibiotics led to the development of a hypothesis and the consequent synthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service