Skip to Content
Merck
All Photos(1)

Documents

S9319

Sigma-Aldrich

Anti-α/β-SNAP (140-157) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

western blot: 1:1,000-1:2,000 using cytosolic fraction S1 of rat and mouse brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

α-SNAP is 80% homologous to β-SNAP and is expressed in all mammalian tissues, whereas β-SNAP is expressed only in brain.
αSNAP (α-soluble N-ethylmaleimide-sensitive fusion (NSF) attachment protein) is widely present and abundantly expressed in all tissues. It is located on human chromosome 19q13.32. βSNAP (β-soluble N-ethylmaleimide-sensitive fusion (NSF) attachment protein) is expressed only in neuronal tissues and is located on human chromosome 20p11.21.

Specificity

Anti-α/β-SNAP (KS-18) recognizes α/β-SNAP.

Immunogen

synthetic peptide encoding amino acids 140-157 located near the mid-region of mouse α-SNAP, conjugated to KLH. The sequence is identical in human, rat, and bovine α-SNAP and in human, mouse, rat and bovine β-SNAP.

Application

Anti-α/β-SNAP (140-157) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

α-SNAP binding to the soluble NSF attachment protein receptor (SNARE) complex, recruits NSF to form a 20S complex. ATP hydrolysis by NSF ATPase activity induces disassembly of the 20S complex, thus facilitating vesicle fusion to the membrane and exocytosis.
α-soluble N-ethylmaleimide-sensitive fusion (NSF) attachment protein (α-SNAP) is an essential constituent of the membrane fusion machinery. It plays an important role in insulin exocytosis through large dense core vesicles. In human epithelia, absence of αSNAP promotes non-canonical autophagy. α-SNAP and NSF are highly essential in the initial stage of human sperm acrosome reaction.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntaxin 13 mediates cycling of plasma membrane proteins via tubulovesicular recycling endosomes.
Prekeris R, et al.
The Journal of Cell Biology, 143(4), 957-971 (1998)
α-Soluble N-ethylmaleimide-sensitive factor attachment protein is expressed in pancreatic β cells and functions in insulin but not γ-aminobutyric acid secretion.
Nagamatsu S, et al.
The Journal of Biological Chemistry, 274(12), 8053-8060 (1999)
Loss of a membrane trafficking protein αSNAP induces non-canonical autophagy in human epithelia.
Naydenov NG, et al.
Cell Cycle, 11(24), 4613-4625 (2012)
Molecular cytogenetic characterization of an inherited maternal duplication 20p11. 21p13 associated with a small 20p11. 21 deletion.
D'angelo CS, et al.
American Journal of Medical Genetics. Part A, 152(12), 3197-3202 (2010)
BKV Agnoprotein Interacts with α-Soluble N-Ethylmaleimide-Sensitive Fusion Attachment Protein, and Negatively Influences Transport of VSVG-EGFP.
Johannessen M, et al.
PLoS ONE, 6(9), e24489-e24489 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service