S8817
SB 218795
>98% (HPLC)
Synonym(s):
(−)-(R)-N-(α-Methoxycarbonylbenzyl)-2-phenylquinoline-4-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C25H20N2O3
Molecular Weight:
396.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Quality Level
Assay
>98% (HPLC)
form
solid
color
white
solubility
DMSO: soluble 40 mg/mL
H2O: insoluble
storage temp.
2-8°C
SMILES string
COC(=O)[C@H](NC(=O)c1cc(nc2ccccc12)-c3ccccc3)c4ccccc4
Gene Information
human ... TACR2(6865) , TACR3(6870)
Application
The pharmacodynamics profile of SB 218795 enables its use as a tool to study physiological and pathophysiological roles of NK-3 receptor in CNS-modulated behaviors.
Biochem/physiol Actions
Non-peptide NK3 tachykinin receptor antagonist.
SB 218795 is a 2-phenyl-4-quinolinecarboxamides and a selective, reversible and competitive antagonist of human NK-3 receptor that effectively crosses the blood-brain barrier. It inhibits the NK-3 receptor-induced miosis or pupil constriction in conscious rabbits.1,2
Legal Information
Sold for research purposes under agreement from GlaxoSmithKline
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A D Medhurst et al.
British journal of pharmacology, 122(3), 469-476 (1997-11-14)
1. Inhibition of NK3 receptor agonist-induced contraction in the rabbit isolated iris sphincter muscle was used to assess the in vitro functional activity of three 2-phenyl-4-quinolinecarboxamides, members of a novel class of potent and selective non-peptide NK3 receptor antagonists. In
H M Sarau et al.
The Journal of pharmacology and experimental therapeutics, 295(1), 373-381 (2000-09-19)
The pharmacological and pharmacokinetic profile of SB-222200 [(S)-(-)-N-(alpha-ethylbenzyl)-3-methyl-2-phenylquinoline-4-car boxami de], a human NK-3 receptor (hNK-3R) antagonist, was determined. SB-222200 inhibited (125)I-[MePhe(7)]neurokinin B (NKB) binding to Chinese hamster ovary (CHO) cell membranes stably expressing the hNK-3 receptor (CHO-hNK-3R) with a K(i)
Jin Zhu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 34(3), 768-780 (2014-08-30)
Activated microglia perform many of the immune effector functions typically associated with macrophages. However, the regulators involved in microglial activation are not well defined. Because microglia play a pivotal role in immune surveillance of the CNS, we studied the effect
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service