Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
UNSPSC Code:
12352203
MDL number:
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
clone
polyclonal
form
lyophilized powder
species reactivity
sheep
packaging
vial of 2 mL lyophilized antiserum
technique(s)
immunoelectrophoresis: suitable
storage temp.
2-8°C
Immunogen
sheep red blood cell stroma
Biochem/physiol Actions
At working dilution 1 mL of antiserum is sufficient to sensitize 1 × 109 sheep red blood cells.
Physical form
Lyophilized from 0.01 M phosphate buffered saline, pH 7.2
Preparation Note
Reconstitute with 2 ml deionized water. Working dilution: 1:100.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Volkan Seyrantepe et al.
The Journal of biological chemistry, 285(1), 206-215 (2009-11-06)
The differentiation of monocytes into macrophages and dendritic cells is accompanied by induction of cell-surface neuraminidase 1 (Neu1) and cathepsin A (CathA), the latter forming a complex with and activating Neu1. To clarify the biological importance of this phenomenon we
Jordan E Inglis et al.
Advances in physiology education, 32(4), 317-321 (2008-12-03)
The immune system is a vital physiological component that affords animals protection from disease and is composed of innate and adaptive mechanisms that rely on cellular and dissolved components. The serum complement system is a series of dissolved proteins that
Related Content
Datasheet
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service