Skip to Content
Merck

Skip To

S7939

Solvent Safe pipette tips

volume range 1-200 μL

Synonym(s):

carbon fiber pipette tips, solvent safe pipette tips, MBP 5069B, dispensing tip, liquid transfer tip

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NB.01
UNSPSC Code:
41121601
Volume:
1—200 μL
Sterility:
non-sterile
Feature:
barrier
Packaging:
pack of 2500 ea (Bag)
Manufacturer/tradename:
Molecular BioProducts 5069B


material

carbon fiber , colorless

sterility

non-sterile

feature

barrier

packaging

pack of 2500 ea (Bag)

manufacturer/tradename

Molecular BioProducts 5069B

L

2 5/16 in. (58.8 mm)

maximum volume

200 μL

volume range

1-200 μL

volume

1—200 μL

compatibility

for use with

  • Biohit Variable 50-200 μl
  • Eppendorf Response 4850 20-500 μl
  • Eppendorf Research/Reference 20-200 μl
  • Eppendorf Research Pro 4860 20-300 μl
  • Finnpipette 4027/4500 Digital 40-200 μl
  • Nichiryo BenchMate 20-200 μl
  • Pipetman P-200 20-200 μl
  • Rainin EDP-Series 250 (Limited Draw)
  • Transferpette 20-200 μl

General description

Solvent Safepipette rips are suitable for use with acids, bases and aggressive organic solvents Revolutionary filter material handles the pipetting challenges of combinatorial chemistry. The unique use of the ART™ filter with pleated activated carbon provides protection for your samples and pipettors against destructive carryover aerosols and vapors from volatile organic solvents without compromising pipetting accuracy and precision.

Application

  • Peptide synthesis
  • HPLC analysis

Features and Benefits

  • Solvent Safe carbon filter pipette tips
  • Uses the new ART filter tip formula (aerosol-resistant tip)
  • Prevents pipetting errors and critical inaccuracies caused by strong acids, bases and aggressive organic solvents
  • A proprietary blend of pleated activated carbon with a 40-micron polyethylene filter provides effective solvent protection
  • Filters block corrosive vapors created when pipetting volatile organic solvents such as TFA
  • Safely and accurately pipette phenol/chloroform DNA extractions
  • Perform peptide synthesis and HPLC analysis
  • Excellent alternative to glass pipettes

Other Notes

Browse all pipette tips

Legal Information

Solvent Safe is a trademark of Molecular BioProducts, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S7814Z740095S8064
sterility

non-sterile

sterility

sterile

sterility

non-sterile

sterility

sterile

manufacturer/tradename

Molecular BioProducts 5069B

manufacturer/tradename

Molecular BioProducts 5469

manufacturer/tradename

BRAND 732032

manufacturer/tradename

Molecular BioProducts 5479E

volume

1—200 μL

volume

1—200 μL

volume

50-1000 μL

volume

100—1000 μL

feature

barrier

feature

barrier

feature

DNase free, RNase and DNase free, RNase free, endotoxin free

feature

barrier

material

carbon fiber , colorless

material

carbon fiber , colorless

material

blue tip, polypropylene , polypropylene tip

material

carbon fiber , colorless

packaging

pack of 2500 ea (Bag)

packaging

pack of 384 ea (4 trays of 96 ea)

packaging

pack of 5000 ea (10 bags of 500)

packaging

pack of 400 ea (4 trays of 100 ea)


Still not finding the right product?

Explore all of our products under Solvent Safe pipette tips




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library