Skip to Content
Merck
All Photos(1)

Key Documents

S6698

Sigma-Aldrich

SRPK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

SFSK1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€494.00

€494.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€494.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€494.00


Estimated to ship on29 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

173-234 nmol/min·mg

mol wt

~130 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SRPK1(6732)

Biochem/physiol Actions

SRPK1 is a Ser/Thr protein kinase that has sequence and function similar to SRPK2. In human cancer cells, it was observed that inactivation of SRPK1 induces cellular resistance to anticancer drugs such as cisplatin and bleomycin. SRPK1 phosphorylates serine/arginine-rich (SR) proteins, such as splicing factors ASF/SF2, SC35, and SRp55, in their arginine/serine-rich (RS) domains in vitro. Hence it is believed that SRPK1 plays a key role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory M Hayes et al.
Cancer research, 67(5), 2072-2080 (2007-03-03)
Aberrant patterns of pre-mRNA processing are typical of human malignancies, yet the mechanisms responsible for these changes remain undefined. We have recently shown overexpression of a core splice regulatory protein, serine-arginine protein kinase 1 (SRPK1), in dysplastic and neoplastic pancreatic
J F Gui et al.
Nature, 369(6482), 678-682 (1994-06-23)
Small nuclear ribonucleoprotein particles (snRNPs) and non-snRNP splicing factors containing a serine/arginine-rich domain (SR proteins) concentrate in 'speckles' in the nucleus of interphase cells. It is believed that nuclear speckles act as storage sites for splicing factors while splicing occurs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service