Skip to Content
Merck
All Photos(1)

Key Documents

S6439

Sigma-Aldrich

SQI-ET sodium form

≥90% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H51N4O7
Molecular Weight:
675.83
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

≥90% (TLC)

storage temp.

−20°C

SMILES string

CCOc1cc(N2CCOCCOCCN(CCOCC2)c3cc(OCC)c(OCC)c4ccc(C)nc34)c5nc(C)ccc5c1OCC

InChI

1S/C38H52N4O7/c1-7-46-33-25-31(35-29(37(33)48-9-3)13-11-27(5)39-35)41-15-19-43-20-16-42(18-22-45-24-23-44-21-17-41)32-26-34(47-8-2)38(49-10-4)30-14-12-28(6)40-36(30)32/h11-14,25-26H,7-10,15-24H2,1-6H3

InChI key

HFXIXJYCUKUTIL-UHFFFAOYSA-N

Application

Sodium ionophore with a selectivity of 100:1 for sodium over potassium. Effective at concentrations of approx. 40 μM.

Physical form

Orange-brown gum

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service