Skip to Content
Merck
All Photos(1)

Key Documents

S5951

Sigma-Aldrich

SW02

Synonym(s):

4-(4-Bromophenyl)-5-(ethoxycarbonyl)-3,4-dihydro-6-methyl-2-oxo-1(2H)-pyrimidinebutanoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€133.00
25 MG
€377.00

€133.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€133.00
25 MG
€377.00

About This Item

Empirical Formula (Hill Notation):
C19H23BrN2O5
CAS Number:
Molecular Weight:
439.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€133.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CCOC(=O)C1=C(C)N(CCCC(=O)OC)C(=O)NC1c2ccc(Br)cc2

InChI

1S/C19H23BrN2O5/c1-4-27-18(24)16-12(2)22(11-5-6-15(23)26-3)19(25)21-17(16)13-7-9-14(20)10-8-13/h7-10,17H,4-6,11H2,1-3H3,(H,21,25)

InChI key

YFZRMRYVWWUQME-UHFFFAOYSA-N

Application

SW02 may be used in Hsp70-mediated cell signaling studies.

Biochem/physiol Actions

SW02 is a specific activator/agonist of Hsp70.
SW02 is a specific activator/agonist of Hsp70. Hsp70 is of particular interest in diseases caused by protein misfolding, such as tau and Aβ in Alzheimer′s. SW02 increases Hsp70 ATPase activity, resulting in increased tau protein but not other disease-related proteins in cells. Hsp70 prevents aggregation of Aβ peptide, as shown by treatment with SW02.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rati Lama et al.
The Journal of steroid biochemistry and molecular biology, 168, 91-101 (2017-02-19)
Hexamethylene bis-acetamide inducible protein 1 (HEXIM1) is identified as a novel inhibitor of estrogen stimulated breast cell growth, and it suppresses estrogen receptor-α transcriptional activity. HEXIM1 protein level has been found to be downregulated by estrogens. Recently, HEXIM1 has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service