Skip to Content
Merck
All Photos(1)

Key Documents

S4199

Millipore

Seppro® Dilution buffer

Synonym(s):

Tris (hydroxymethyl) aminomethane

Sign Into View Organizational & Contract Pricing

Select a Size

200 ML
€124.00

€124.00


Check Cart for Availability


Select a Size

Change View
200 ML
€124.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.32

€124.00


Check Cart for Availability

storage temp.

2-8°C

Application

Seppro® Dilution buffer [Tris (hydroxymethyl) aminomethane] is a reagent qualified for use in Seppro® protein preparation and separation systems.[1]
Seppro® Dilution buffer has been used as a dilution buffer for IgY immunodepletion studies.[2][3]

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerard Such-Sanmartín et al.
Analytical chemistry, 86(3), 1543-1550 (2014-01-17)
Protein and proteome analysis of human blood plasma presents a challenge to current analytical platforms such as mass spectrometry (MS). High abundance plasma proteins interfere with detection of potential protein biomarkers that are often 3-10 orders of magnitude lower in
Effect of plasma protein depletion on BNP-32 recovery.
Adam M Hawkridge et al.
Clinical chemistry, 54(5), 933-934 (2008-04-30)
Simon Sheng et al.
Methods in molecular biology (Clifton, N.J.), 728, 29-46 (2011-04-07)
Serum- and plasma-based biomarker discovery requires technologies with specific capabilities: sufficient proteome coverage and depth, technical reproducibly, and the scalability to enable analysis on a large number of samples at reasonable cost. We have shown that plasma samples processed using
Hasmik Keshishian et al.
Nature protocols, 12(8), 1683-1701 (2017-07-28)
Proteomic characterization of blood plasma is of central importance to clinical proteomics and particularly to biomarker discovery studies. The vast dynamic range and high complexity of the plasma proteome have, however, proven to be serious challenges and have often led

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service